missing translation for 'onlineSavingsMsg'
Learn More

2,4-pentanedionato de holmio(III), REacton™, 99,9 % (REO), Thermo Scientific Chemicals

Catalog Number p-7049202
Click to view available options
Cantidad:
2 g
10 g
This item is not returnable. View return policy
14589-33-4
C15H21HoO6
462.26
MFCD00050176
UEKRGRZSLATUQV-UHFFFAOYSA-N
holmium 2,4-pentanedionate, holmium iii 2,4-pentanedionate, reacton, holmium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy holmio oxy pent-3-en-2-one
24282829
holmium(3+) tris(2,4-dioxopentan-3-ide)
[Ho+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
This item is not returnable. View return policy
14589-33-4
C15H21HoO6
462.26
MFCD00050176
UEKRGRZSLATUQV-UHFFFAOYSA-N
holmium 2,4-pentanedionate, holmium iii 2,4-pentanedionate, reacton, holmium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy holmio oxy pent-3-en-2-one
24282829
holmium(3+) tris(2,4-dioxopentan-3-ide)
[Ho+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

It is used for Building Blocks of chemicals and , Chemical Synthesis, used as a trihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Holmium(III) 2,4-pentanedionate
Forma física Polvo
Cantidad 2 g
Notas del porcentaje del ensayo (REO)
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 462.26
Porcentaje de pureza 99.9%
Grado Reacton™
Olor Odorless
Sensibilidad Hygroscopic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'