missing translation for 'onlineSavingsMsg'
Learn More

Lámina de holmio, 0,3 mm de grosor, 99,9 % (REO), Thermo Scientific Chemicals

Catalog Number p-7048343
Click to view available options
Cantidad:
25 x 25 mm
50 x 50 mm
Este artículo no se puede devolver. Vea la política de devoluciones
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
23988
CHEBI:49648
holmio
[Ho]
Este artículo no se puede devolver. Vea la política de devoluciones
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
23988
CHEBI:49648
holmio
[Ho]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Envasado bajo argón
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Holmium foil
Nombre de nota 0.3mm (0.01 in.) thick
Notas del porcentaje del ensayo (REO)
Cantidad 25 x 25 mm
Porcentaje de pureza 99.9%
Olor Odorless
Envase Mylar bajo argón
Peso ≈1.65g/25x25mm
Formulario Papel de aluminio

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'