Learn More
Hoefer™ SE 600 Series Complete Vertical Electrophoresis System
Product Code. 10321575
Change view
Click to view available options
Voltage:
1000 V
Unit Size:
Each
Product Code. Voltage Unit Size
10321575 1000 V Each
1 options
This item is not returnable. View return policy
Product Code. 10321575 Supplier Hoefer™ Supplier No. SE600151.5
  • ONLINE EXCLUSIVE PRICE
    1960.00€
    1912.04€ / paquete de 1
    Ahorro 47.96€
    2% Descuento
View all promotions
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Built-in heat exchanger maintains temperature between 1° and 45°C (33.8° and 113°F) for faster runs with superior quality results
  • Offers durable construction for proven decades of performance
  • Run one gel or two gels simultaneously, four with optional divider plates
  • Produce straight lanes and sharp, well-defined bands—no “smiling”
  • Provide two mechanisms to dissipate heat
  • Gel sandwiches are completely immersed in the lower buffer so that heat is transferred evenly on both sides of the gel
  • A heat exchanger suspended vertically in the lower chamber cools or heats buffer evenly
Suitable for denaturing and native polyacrylamide, 2-D, isoelectric focusing and agarose gel electrophoresis
Certifications
UL3101-1, CSA C22.2 1010.1, CE Certified, EN61010-1
Specifications
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Humidity Range Up to 80%
Includes Lower Buffer chamber, upper Buffer chamber, safety lid with high voltage Leads, heat exchanger, Glass plates, clamp assemblies, cams, slotted gaskets for upper Buffer chamber, dual gel casting stand with leveling base, bubble level, laminated gaskets for
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15°C to 30°C
Thickness (English) Spacer 0.05 in.
Thickness (Metric) Spacer 0.15 cm
Volume (English) Buffer 169.07 oz.
Volume (Metric) Buffer 5000 mL
Width (English) 12.59 in.
Width (English) Gel 7.08 in.
Width (Metric) 32 cm
Width (Metric) Gel 18 cm
Height (English) 11.41 in.
Height (English) Gel 6.29/3.14 in.
Height (Metric) 29 cm
Voltage 1000 V
Wattage 50 w
Amperage 500 mA