Learn More
Hoefer™ HE 99X Max Submarine Electrophoresis Unit
Product Code. 12004817
Change view
Click to view available options
Model:
HE 99X Max Basic
HE 99X Max Complete
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
12004817 HE 99X Max Complete Each
11519573 HE 99X Max Basic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12004817 Supplier Hoefer™ Supplier No. HE99X151.5
  • ONLINE EXCLUSIVE PRICE
    800.00€
    703.49€ / paquete de 1
    Ahorro 96.51€
    12% Descuento
View all promotions
Fecha estimada de envoi: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Includes
Buffer chamber with electrodes and leveling feet, safety lid with leads, bubble level. Complete system (03-500-121) also includes 15 x 20cm gel casting tray and UV-transparent running tray; 1.5mm thick, 15-well comb; adjustable comb back that permits control of the depth of the sample wells
Offers no-mess gel casting without tape or dams
  • Unit accommodates a variety of gel lengths for maximum resolution of DNA fragments
  • Agarose bridges are also accommodated, so gels can be run in nonsubmarine fashion
  • Rugged design provides reliability and safety
Specifications
Certifications/Compliance EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Type Horizontal Agarose Electrophoresis Unit
Humidity Range Up to 80%
Includes Buffer chamber assembly, safety lid with high voltage Leads, 15 x 20cm running tray, 15 x 20cm casting tray, Foam gaskets, bubble level, 1.5mm thick, 15-well comb, Comb Back
Model HE 99X Max Complete
Sample Size 60
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (English) Buffer 40.5 oz.
Volume (Metric) Buffer 1200 mL
Dimensions (L x W x H) 14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm)
Length (English) 14.1 in.
Length (English) Gel 5.9, 3.9 or 7.8 in.
Length (Metric) 36 cm
Length (Metric) Gel 15, 10 or 20 cm
Width (English) 7.1 in.
Width (English) Gel 5.9 in.
Width (Metric) 18.2 cm
Width (Metric) Gel 15 cm
Height (English) 5.5 in.
Height (Metric) 14 cm
Voltage 200 V
Wattage 20 w
Amperage 100 mA