Learn More
HLA G Antibody (G233), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB110-55298UV
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214053
  • 532.38€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for Flow Cytometry, ELISA.
Especificaciones
Especificaciones
HLA G
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
HLA-G
HLA-A2.1/human beta2-microglobulin double transgenic mice were immunized with murine L cells transfected with both human beta2-microglobulin and HLA-G.
0.1 mL
Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
3135
Human
Purified
Flow Cytometry, ELISA
G233
Flow Cytometry, ELISA
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G
Mouse
Protein A purified
RUO
Primary
This antibody (clone G233) recognizes several isoforms of HLA-G expressed in all populations of extravillous trophoblast (cell columns, interstitial trophoblast, endovascular trophoblast, placental bed giant cells). HLA-G belongs to the nonclassical MHC Class I molecules (MHC Class Ib). The antibody has been found not to cross-react with any other MHC Class I antigens (HLA-A, -B, -C, -E, -F).
Store at 4C in the dark.
IgG2a