Learn More
HLA-E Antibody (MEM-E/07), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB500-310R
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214043
  • 497.83€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
HLA-E Monoclonal specifically detects HLA-E in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
HLA-E
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
HLA-E
Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide.
0.1 mL
Adaptive Immunity, Asthma, Diabetes Research, Immunology, Major Histocompatibility Complexes
3133
Human
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
MEM-E/07
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Mouse
Protein A purified
RUO
Primary
The antibody MEM-E/07 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that the antibody cross-reacts with some classical MHC Class I molecules (MHC Class Ia): HLA-B7 (strongly), HLA-B8 (moderately), HLA-B27, -B44 (weakly).
Store at 4C in the dark.
IgG1