missing translation for 'onlineSavingsMsg'
Learn More

HLA DQ1/DQ3 Antibody (HL-37), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NB500-405IR

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214440

  • 497.83€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

HLA DQ1/DQ3 Monoclonal specifically detects HLA DQ1/DQ3 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones

Especificaciones

HLA DQ/DQ3
Monoclonal
DyLight 755
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Mouse
Affinity Purified
RUO
Primary
The antibody HL-37 reacts with polymorphic determinant on human HLA-DQ1 and HLA-DQ3 molecules (recognized epitope was found on isolated beta chain of DQ1), but does not react with HLA-DQ2. DQ is the isotype of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages).
Store at 4C in the dark.
Western Blot, Flow Cytometry
HL-37
Western Blot, Flow Cytometry
HLA-DQA1
Burkitt's lymphoma cell line Raj
0.1 mL
Adaptive Immunity, Asthma, Immunology
3117
Human, Porcine
IgG3
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only