missing translation for 'onlineSavingsMsg'
Learn More

HLA-ABC Mouse anti-Human, Super Bright 600, Clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 63-9983-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923584

  • 133.00€ / paquete de 25


Fecha estimada de envío
20-08-2020
Añadir a la cesta

Descripción

Descripción

Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. HLA-class I antigens are widely distributed on human nucleated cells. The intensity of HLA-ABC maybe altered in pathological states e.g. maligt cells may lose HLA-ABC, whereas hepatocytes in alcoholic hepatitis, biliary cirrhosis and chronic active hepatitis may show enhanced expression of HLA-ABC.
Especificaciones

Especificaciones

HLA-ABC
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P04439
H-2K, H-2K(d), H2-K, MGC184092, MGC7052
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
W6/32
Super Bright 600
HLA-A
Liquid
HLA-A
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3105
Videos
Documentos