missing translation for 'onlineSavingsMsg'
Learn More

HLA-ABC Mouse anti-Human, Super Bright 436, Clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 62-9983-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933454

  • 139.00€ / paquete de 25


Fecha de envío estimada
del proveedor 31-01-2020
Añadir a la cesta

Descripción

Descripción

Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. HLA-class I antigens are widely distributed on human nucleated cells. The intensity of HLA-ABC maybe altered in pathological states e.g. maligt cells may lose HLA-ABC, whereas hepatocytes in alcoholic hepatitis, biliary cirrhosis and chronic active hepatitis may show enhanced expression of HLA-ABC.
Especificaciones

Especificaciones

HLA-ABC
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P05534
HLA-A
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3105
Flow Cytometry
W6/32
Super Bright 436
HLA-A
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
Documentos