Learn More
HLA A2 Antibody (BB7.2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP1-44896AF488
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216211
  • 491.35€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
HLA A2 Monoclonal specifically detects HLA A2 in Human samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
HLA A2
Monoclonal
Alexa Fluor 488
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Mouse
Protein A or G purified
RUO
Primary
The antibody BB7.2 recognizes an epitope at the C-terminus of alpha-2 helix and a turn on one of the underlying beta strands within the human HLA-A2 histocompatibility antigen.
Store at 4C in the dark.
IgG2b
Flow Cytometry
BB7.2
Flow Cytometry
HLA-A
HLA-A2 solubilised by papain
0.1 mL
Adaptive Immunity, Cell Biology, Immunology
3105
Human
Purified