Learn More
Epredia™ HistoScreen™ Cassettes in Tube Packs
Product Code. 16303946 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16303946 Lilac Pack of 750
16343936 Beige Pack of 750
16363936 Green Pack of 750
16373936 Gray Pack of 750
16383936 Lavender Pack of 750
16393936 Light Green Pack of 750
16313946 Orange Pack of 750
16323946 Pink Pack of 750
16333946 Tan Pack of 750
16343946 White Pack of 750
16353946 Yellow Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 16303946 Supplier Epredia™ Supplier No. B851001LI
  • ONLINE EXCLUSIVE PRICE
    450.00€
    350.70€ / paquete de 750
    Ahorro 99.30€
    22% Descuento
 Restricted Item
Fecha estimada de envoi: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Eliminates the need for sponges or bags, ideal for very small biopsy tissue samples. Cassettes are compatible with Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
Color Lilac
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type HistoScreen Cassette