Learn More
Histone H4, ac Lys5 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NB21-2024AF700
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213840
  • 663.05€ / 0.05 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Porcine, Mammal samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
Histone H4 (ac Lys5)
Polyclonal
Western Blot, Immunocytochemistry/Immunofluorescence
P62805
HIST4H4
Synthetic aceylated peptide surrounding Lysine 5 of human Histone H4 [Swiss Prot P62805].
0.05 mL
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
121504
Human, Mouse, Rat, Porcine, Mammalia
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Rabbit
Affinity Purified
RUO
Primary
Human, mouse and rat. Predicted to react with most mammal species.
Store at 4C in the dark.