missing translation for 'onlineSavingsMsg'
Learn More

His Tag Antibody (3D5), DyLight 650, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Marca:  Novus Biologicals NBP2-81022C

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213429

  • 595.02€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

His Tag
Monoclonal
DyLight 650
50mM Sodium Borate
Armenian Hamster
Protein A purified
RUO
Primary
Store at 4°C in the dark.
IgG κ
Western Blot, ELISA
3D5
Western Blot, ELISA
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Purified his tagged Fab fragment.
0.1 mL
Cellular Markers, Epitope Tags
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags.
Purified
Epitope Tag
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales