Learn More
HIRIP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 14992-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16813954
  • 319.00€ / 150 microlitros
Envío estimado: 27-10-2021
Descripción
Descripción
The HIRA protein shares sequence similarity with Hir1p and Hir2p, the two corepressors of histone gene transcription characterized in the yeast, Saccharomyces cerevisiae. The structural features of the HIRA protein suggest that it may function as part of a multiprotein complex. Several cDNAs encoding HIRA-interacting proteins, or HIRIPs, have been identified. In vitro, the protein encoded by this gene binds HIRA, as well as H2B and H3 core histones, indicating that a complex containing HIRA-HIRIP3 could function in some aspects of chromatin and histone metabolism. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Especificaciones
Especificaciones
HIRIP3
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BLH7, Q9BW71
HIRA interacting protein 3, HIRIP3
Rabbit
IgG
150 μL
-20°C
Primary
233876, 8479
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HIRIP3
Liquid
HIRIP3
HIRIP3 Fusion Protein Ag6978
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse