Learn More
Invitrogen™ HIPK1 (Myak) Recombinant Human Protein
HIPK1 phosphorylates homeodomain transcription factors.
Marca: Invitrogen™ PR8290A
Detalles adicionales : Peso : 0.01000kg
Descripción
HIPK1 may also function as a co-repressor for homeodomain transcription factors.
Especificaciones
NP_689909 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
HIPK1 (Myak) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
73.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |