Learn More
Corning™ High Base Cyclic Olefin Copolymer Microplate
Raised well bottoms for higher sensitivity. The High Base Cyclic Olefin Copolymer Microplate has excellent signal dynamic range.
Marca:  Corning™ 4370
Detalles adicionales : Peso : 9.00000kg
 Ver más versiones de este producto
Código de producto. 15823885
  • 1056.15€ / paquete de 50
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Raised well bottoms for higher sensitivity
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
  • Barcoded
Especificación
Especificación
Yes
White
1536
12.5μL
Not Included
TC Treated
Flat
Cyclic Olefin Copolymer
50
Up to 8μL
10
Sterile
Videos