missing translation for 'onlineSavingsMsg'
Learn More

Hexokinase 2 Antibody (1A7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-04296V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215393

  • 558.30€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

Hexokinase 2
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
HK2
Recombinant human Hexokinase 2 (1-917aa) purified from E. coli
0.1 mL
Cell Cycle and Replication
3099
Human
Purified
Western Blot, ELISA
1A7
Western Blot, ELISA
DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase
Mouse
Protein G purified
RUO
Primary
Anti-human Hexokinase 2 mAb, clone 1A7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Hexokinase 2 protein.
Store at 4C in the dark.
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales