Learn More
Abnova™ Hepatitis B surface antigen Ab ELISA Kit
Sandwich Enzyme Immunoassay for the measurement of antibodies to Hepatitis B surface antigen
Marca: Abnova™ KA0286.1Kit
Detalles adicionales : Peso : 0.02000kg
Especificaciones
Hepatitis B surface antigen ELISA Kit is a sandwich enzyme immunoassay for the measurement of hepatitis B surface antigen. | |
Qualitative | |
Hepatitis B surface antigen ELISA Kit | |
Plasma, Serum | |
Store the kit at 4°C. |
Colorimetric | |
Hepatitis B surface antigen ELISA Kit | |
1 Kit | |
50μL |