missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemopexin Mouse anti-Human, Porcine, Rat, Clone: 3A9D6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66479-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.Especificaciones
Hemopexin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02790, P20059, P50828 | |
HPX | |
Hemopexin Fusion Protein Ag8533 | |
150 μL | |
Primary | |
Human, Porcine, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3A9D6 | |
Unconjugated | |
HPX | |
Beta 1B glycoprotein, hemopexin, HPX | |
Mouse | |
Protein G | |
RUO | |
3263, 396998, 58917 | |
-20°C | |
Liquid |