missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEK 293 cells Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27347-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is designed to detect the protein of the HEK293 cells.
Human Embryonic Kidney 293 cells, also often referred to as HEK 293, 293 cells, or less precisely as HEK cells are a specific cell line originally derived from human embryonic kidney cells grown in tissue culture. HEK 293 cells were generated in early 70s by transformation of cultures of normal human embryonic kidney cells with sheared adenovirus 5 DNA in Alex Van der Eb′s laboratory in Leiden, Holland.Especificaciones
HEK 293 cells | |
Polyclonal | |
Unconjugated | |
293, HEK 293 cells, HEK293 | |
Peptide | |
150 μL | |
Primary | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG |