missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEATR2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24578-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is essential for the preassembly or stability of axonemal dynein arms, and is found only in organisms with motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia-18, a disorder characterized by abnormalities of motile cilia. Alternatively spliced transcript variants have been found for this gene.Especificaciones
HEATR2 | |
Polyclonal | |
Unconjugated | |
DNAAF5 | |
HEAT repeat containing 2, HEATR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304332, 54919 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86Y56 | |
DNAAF5 | |
HEATR2 Fusion Protein Ag20080 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |