Learn More
Bibby Scientific™ Heating Element for Soxhlet
Product Code. 11826215
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
11826215 250 mL Each
1 options
This item is not returnable. View return policy
Product Code. 11826215 Supplier Bibby Scientific™ Supplier No. REME0250
  • ONLINE EXCLUSIVE PRICE
    221.60€
    196.00€ / paquete de 1
    Ahorro 25.60€
    11% Descuento
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Bibby Scientific Heating Element for Soxhlet, Capacity: 250 mL, For Use With: Spare Soxhlet Extractor
Specifications
Capacity (Metric) 250 mL
For Use With Spare Soxhlet Extractor
Product Type Heating Element