Learn More
Huenersdorf™ HDPE Industrial Bucket
Product Code. 11863652
Change view
Click to view available options
Color:
Natural
Description:
Round
Square
Unit Size:
Each
Capacity (Metric):
10.5 L
17 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Description Unit Size Capacity (Metric)
11863652 Natural Square Each -
11796715 Natural Round Each 17 L
11816124 Natural Round Each 10.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11863652 Supplier Huenersdorf™ Supplier No. 943100
  • ONLINE EXCLUSIVE PRICE
    43.05€
    13.03€ / paquete de 1
    Ahorro 30.02€
    69% Descuento
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Use the Huenersdorf™ HDPE Industrial Bucket for all your laboratory needs. It is food-safe and has a spout for safe handling.
  • Round with spout
  • Natural
  • Acid-resistant polyethylene (HDPE)
  • Embossed scale and plastic handle Industrial bucket with spout for safe handling
Specifications
Product Type Industrial Bucket
Material HDPE
Color Natural
Description Square
Dimensions 292 x 292 x 292 mm
Height (Metric) 292 mm
Includes w/out Spout
Length (Metric) 292 mm