Learn More
Huenersdorf™ HDPE Industrial Bucket
Product Code. 11816124
Change view
Click to view available options
Capacity (Metric):
10.5 L
17 L
Color:
Natural
Description:
Round
Square
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Description Unit Size
11816124 10.5 L Natural Round Each
11796715 17 L Natural Round Each
11863652 - Natural Square Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11816124 Supplier Huenersdorf™ Supplier No. 941100
  • ONLINE EXCLUSIVE PRICE
    34.25€
    9.03€ / paquete de 1
    Ahorro 25.22€
    73% Descuento
En stock
Add to basket
Edge
Add to basket
Use the Huenersdorf™ HDPE Industrial Bucket for all your laboratory needs. It is food-safe and has a spout for safe handling.
  • Round with spout
  • Natural
  • Acid-resistant polyethylene (HDPE)
  • Embossed scale and plastic handle Industrial bucket with spout for safe handling
Specifications
Product Type Industrial Bucket
Capacity (Metric) 10.5 L
Material HDPE
Color Natural
Description Round
Diameter (Metric) Outer 286 mm
Height (Metric) 283 mm
Includes Spout
Quantity 1 Ea.