missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems HCoV-NL63 S1 protein, His Tag

3181.91€

Especificaciones

Formato Powder
Peso molecular 79.5 kDa
Pureza >90% as determined by SDS-PAGE
Fuente HCoV-NL63 S1 protein, His Tag (SIN-V52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 19 - Val 717 (Accession # Q6Q1S2-1).
Inmunógeno Cys 19 - Val 717
Ver más especificaciones

Productos 1
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17186711
Ver documentos Detalles de la promoción
ACROBiosystems
SIN-V52H3-1MG
1 mg
3181.91€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 09-01-2023
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.5 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Especificaciones

Especificaciones

Powder
>90% as determined by SDS-PAGE
Cys 19 - Val 717
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
PBS, pH7.4
HEK293
HCoV-NL63
79.5 kDa
HCoV-NL63 S1 protein, His Tag (SIN-V52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 19 - Val 717 (Accession # Q6Q1S2-1).
-20°C
S1 protein
His Tag
Less than 1.0 EU / µg by the LAL method.
Viral Proteins
SDS
Product Certifications