missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems HCoV-NL63 S1 protein, His Tag

Marca:  ACROBiosystems SIN-V52H3-1mg

Detalles adicionales : Peso : 0.00010kg

Código de producto. 17186711

  • 3411.22€ / 1 miligramo
Fecha estimada de envío: 30-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.5 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Especificaciones

Especificaciones

79.5 kDa
HCoV-NL63 S1 protein, His Tag (SIN-V52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 19 - Val 717 (Accession # Q6Q1S2-1).
-20°C
S1 protein
HCoV-NL63
PBS, pH7.4
HEK293
Viral Proteins
1 mg
Cys 19 - Val 717
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales