Learn More
3M™ Hazardous Spill Response Kit
Product Code. 17119350
Change view
Click to view available options
Dimensions (L x W):
1040 x 640 mm
Unit Size:
1 set
Product Code. Dimensions (L x W) Unit Size
17119350 1040 x 640 mm 1 set
1 options
This item is not returnable. View return policy
Product Code. 17119350 Supplier 3M™ Supplier No. 7100090072
  • ONLINE EXCLUSIVE PRICE
    4065.00€
    2571.59€ / 1 sets
    Ahorro 1493.41€
    36% Descuento
 Restricted Item
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Ensures that all necessary products are easily accessible should a spill occur. Quick clean up of hazardous chemical spills is crucial in an emergency situation. Includes a selection of 3M™ Chemical Sorbents in various formats, in addition to clean up bags.
  • Ideal for rapid response clean up of hazardous chemicals and non-aggressive liquid spills
  • Designed to absorb and contain small spills or leaks
  • Fast, effective containment and clean-up
  • Easy to dispose off
TRUSTED_SUSTAINABILITY
Specifications
Type Spill Response Kit
For Use With (Application) Hazardous chemicals and non-aggressive liquid spills
Model HSRK-210
Packaging Quantity 1 Ea.
Dimensions (L x W) 1040 x 640 mm
Length (Metric) 1040 mm
Width (Metric) 640 mm