missing translation for 'onlineSavingsMsg'
Learn More

HARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11301-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818283

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody can bind both HARSs for the close sequences.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes likely share a bidirectional promoter. Mutations in this gene are associated with the pathogenesis of Perrault syndrome, which involves ovarian dysgenesis and sensorineural hearing loss. Alternative splicing results in multiple transcript variants of this gene.
Especificaciones

Especificaciones

HARS2
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49590, Q99KK9
HARS2, HARSL, HARSR, HisRS, Histidine tRNA ligase, Histidine tRNA ligase like, HO3
Rabbit
IgG
150 μL
-20°C
Primary
23438, 307491, 70791
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HARS2
Liquid
Hars2
HARS2 Fusion Protein Ag1827
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados