Learn More
Dihidrato de clorhidrato de Harmol, 98 %, Thermo Scientific Chemicals
Catalog Number p-7042805
Click to view available options
Cantidad:
1 g
5 g
149022-16-2
C12H15ClN2O3
270.713
MFCD00150058
OITPZRQHDRVLMO-UHFFFAOYSA-N
harmol hydrochloride dihydrate, 1-methyl-9h-pyrido 3,4-b indol-7-ol hydrochloride dihydrate, harmol hydrochloride hydrate, harmol dihydrate hydrochloride, c12h10n2o.hcl.2h2o, 1-methyl-2,9-dihydropyrido 3,4-b indol-7-one dihydrate hydrochloride
2723830
1-metil-2,9-dihidropirido[3,4-b]indol-7-ona;dihidrato;clorhidrato
CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1.O.O.Cl
149022-16-2
C12H15ClN2O3
270.713
MFCD00150058
OITPZRQHDRVLMO-UHFFFAOYSA-N
harmol hydrochloride dihydrate, 1-methyl-9h-pyrido 3,4-b indol-7-ol hydrochloride dihydrate, harmol hydrochloride hydrate, harmol dihydrate hydrochloride, c12h10n2o.hcl.2h2o, 1-methyl-2,9-dihydropyrido 3,4-b indol-7-one dihydrate hydrochloride
2723830
1-metil-2,9-dihidropirido[3,4-b]indol-7-ona;dihidrato;clorhidrato
CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1.O.O.Cl
Induce a la apoptosis mediante la activación de la caspasa-8
Harmol hydrochloride dihydrate is used to induces apoptosis by Caspase-8 activation, CNS stimulant, may act through NMDA receptors, and antitumor agent. Harmol has been shown to inhibit haspin kinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El dihidrato de hidrocloruro de Harmol se usa para inducir la apoptosis mediante la activación de la caspasa-8, estimulante del SNC, puede actuar a través de receptores NMDA y agentes antitumorales. Se ha demostrado que Harmol inhibe la quinasa de haspin.
Solubilidad
Soluble en agua.
Notas
Almacenar en un lugar fresco y seco y en recipientes bien sellados. Mantener el recipiente herméticamente cerrado.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Harmol hydrochloride dihydrate
Color Blanco
Cantidad 1 g
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 270.72 (234.69 Anhydrous)
Porcentaje de pureza 98%
Forma física Polvo