Learn More
MP Biomedicals™ Ham's F-10 with L-Glutamine, without Sodium Bicarbonate, Powder
Product Code. 17308593 Shop All MP Biomedicals Products
Change view
Click to view available options
Form:
Powder
Unit Size:
10 L
Product Code. Form Unit Size
17308593 Powder 10 L
1 options
This item is not returnable. View return policy
Product Code. 17308593 Supplier MP Biomedicals™ Supplier No. 091040120
  • ONLINE EXCLUSIVE PRICE
    171.00€
    162.75€ / 10 litros
    Ahorro 8.25€
    4% Descuento
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Ham's F-10 powder with L-Glutamine and without Sodium Bicarbonate, used for cell cultures.
Originally, Ham's Nutrient Mixtures were developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells. Both F-10 and F-12 mediums are formulated for use with or without serum, depending on the type of cells being cultured.
F-10 has demonstrated satisfactory growth of:
  • Human diploid cells
  • White blood cells for chromosomal analysis
  • Primary rat, rabbit and chicken tissue explants
Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
Specifications
Form Powder
Product Type Cell Culture Medium
With Additives Glucose, Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No Sodium Bicarbonate