Learn More
Lipomed Haloperidol-D4, 0.1mg/ml in Methanol
Product Code. 30457479
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30457479 1 mL 1mL
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457479 Supplier Lipomed Supplier No. LPMHLP1902FB0.1LM
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Lipomed's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Haloperidol-D4
CAS 136765-35-0
Solvent or Matrix Methanol
Molecular Formula C21 D4 H19 Cl F N O2
Quantity 1 mL
Synonym Haloperidol-D4
InChI Formula InChI=1 S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26 H,1-2,11-15H2/i3D,4 D,9 D,10 D
SMILES [2 H]c1c([2 H])c(C(=O)CCCN2CCC(O)(CC2)c3ccc(Cl)cc3)c([2 H])c([2 H])c1F
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidyl]-1-(2,3,5,6-tetradeuterio-4-fluoro-phenyl)butan-1-one
Molecular Weight (g/mol) 379.89
Formula Weight 379.1652