missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTCD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17502-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
GSTCD (glutathione S-transferase, C-terminal domain containing) is a 633 amino acid protein belonging to the GSTCD family and contains one GST C-terminal domain. The gene encoding GSTCD maps to human chromosome 4, which represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington′s disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer.Especificaciones
GSTCD | |
Polyclonal | |
Unconjugated | |
GSTCD | |
GSTCD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310855, 67553, 79807 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5RL51, Q8NEC7 | |
Gstcd | |
GSTCD Fusion Protein Ag11690 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |