Learn More
GST (Epitope Tag) Ab-1 Mouse Monoclonal Antibody, 200 μg/mL, BSA and azide,Lab Vision™ Epredia™
Ensure accurate, reproducible results in western blotting and affinity purification applications with Epredia™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-707-P0
Detalles adicionales : Peso : 1.00000kg
Descripción
Host Species: Mouse
Clone: GST01; same as 794
Isotype: IgG1
Species Reactivity: Proteins from all species that are tagged with GST
Immunogen: Purified recombinant GST protein
Cellular Localization: Depends upon the localization of the parent protein tagged with GST
Recommended for:
- Western Blotting
- Affinity Purification
Note:
USA: RUO; Int'l: RUO
Especificaciones
GST (Epitope Tag) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Primary | |
Concentrated |
Western Blot | |
GST01 (794) | |
Unconjugated | |
Purified recombinant GST protein | |
RUO | |
All | |
IgG1 |