missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRPEL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17751-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
GRPEL2 (GrpE Like 2, Mitochondrial) is a Protein Coding gene. Among its related pathways are Mitochondrial protein import and Metabolism of proteins. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL1. Essential component of the PAM complex, a complex required for the translocation of transit peptide-containing proteins from the inner membrane into the mitochondrial matrix in an ATP-dependent manner. Seems to control the nucleotide-dependent binding of mitochondrial HSP70 to substrate proteins. Stimulates ATPase activity of mt-HSP70. May also serve to modulate the interconversion of oligomeric (inactive) and monomeric (active) forms of mt-HSP70.Especificaciones
GRPEL2 | |
Polyclonal | |
Unconjugated | |
GRPEL2 | |
GRPEL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134266, 17714, 688777 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88396, Q8TAA5 | |
Grpel2 | |
GRPEL2 Fusion Protein Ag12018 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |