missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13990-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The product of this gene phosphorylates the beta-2-adrenergic receptor and appears to mediate agonist-specific desensitization observed at high agonist concentrations. This protein is an ubiquitous cytosolic enzyme that specifically phosphorylates the activated form of the beta-adrenergic and related G-protein-coupled receptors. Abnormal coupling of beta-adrenergic receptor to G protein is involved in the pathogenesis of the failing heart.Especificaciones
GRK2 | |
Polyclonal | |
Unconjugated | |
GRK2 | |
ADRBK1, BARK, BARK1, Beta ARK 1, BETA ARK1, GRK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
156 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25098 | |
GRK2 | |
GRK2 Fusion Protein Ag5071 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |