Learn More
Honeywell™ GRIP Latex Gloves
Honeywell™ GRIP Latex Gloves for handling of heavy and slightly sharp parts in a damp or non greasy environment.
Marca:  Honeywell™ 2094145-09
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12655203
  • 6.30€ / cada uno
Stock Remoto 3
Añadir a la cesta
Descripción
Descripción
  • Natural latex coated glove on cotton jersey base
  • Crinkled finishing
  • Knitted wrist
  • The cotton absorbs perspiration thus provides greater comfort
  • The ventilated back improves the glove's breathability
  • The glove's crinkled latex coating provides resistance to abrasion and perforation as well as an excellent grip
  • Natural latex provides excellent flexibility, synonymous with dexterity and comfort
  • It also offers good abrasion and tear resistance
  • Its crepe finish helps the user to grasp slippery parts
Especificaciones
Especificaciones
Latex
Cotton
26 cm
Crinkled
9
EN 420-2003 Basic standard, EN 388-2016 Mechanical risks