Learn More
Hecht Karl™ Graduated Glass Cylinder
Product Code. 10014111
Change view
Click to view available options
Capacity (Metric):
10 mL
100 mL
200 mL
250 mL
1000 mL
Height (Metric):
90 mm
195 mm
250 mm
265 mm
325 mm
440 mm
Description:
Low Form
Tall Form
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Description Unit Size
10014111 200 mL 195 mm Low Form Each
10238391 100 mL 250 mm Tall Form Each
10410762 10 mL 90 mm Low Form Each
10450192 1000 mL 440 mm Tall Form Each
10525762 250 mL 325 mm Tall Form Each
10746952 200 mL 265 mm Tall Form Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10014111 Supplier Hecht Karl™ Supplier No. 44401042
  • ONLINE EXCLUSIVE PRICE
    12.30€
    9.16€ / paquete de 1
    Ahorro 3.14€
    25% Descuento
 Restricted Item
Fecha estimada de envoi: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Material Glass
Capacity (Metric) 200 mL
Description Low Form
Height (Metric) 195 mm
Product Type Graduated Cylinder