Learn More
Sartorius Stedim Biotech™ Grade 601/N Creped Papers for Qualitative and Technical Analyses, Discs
Sartorius Stedim Biotech™ Grade 601/N Creped Papers for Qualitative and Technical Analyses, Discs are suitable for analytical and technical purposes and feature a medium fast flow rate due to their structure.
Marca:  Sartorius Stedim Biotech™ FT-3-354-180
Detalles adicionales : Peso : 0.20000kg
 Ver más versiones de este producto
Código de producto. 15912940
  • 13.05€ / paquete de 100
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Made of refined pulp and cotton linters with > 95% alpha-cellulose
  • Ash content between 0.1 and 0.15%
  • Medium fast filtering
  • Wet-strengthened
  • Non-sterile, bulk-packed
Especificaciones
Especificaciones
100/pk.
180 mm
Cotton Linters and Refined Pulp (> 95 % alpha-cellulose)
12 to 14μm
65g/m2
Qualitative Filter Paper
601/N
0.19 mm
Disc