missing translation for 'onlineSavingsMsg'
Learn More

GPRASP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12159-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16819703

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of a family that regulates the activity of G protein-coupled receptors (GPCRs). The encoded protein has been shown to be capable of interacting with several GPCRs, including the M1 muscarinic acetylcholine receptor and the calcitonin receptor. Several transcript variants encoding the same protein have been found for this gene.
Especificaciones

Especificaciones

GPRASP2
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BUY8, Q96D09
GASP 2, GASP2, GPRASP2
Rabbit
IgG
20 μL
-20°C
Primary
114928, 245607, 317405
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GPRASP2
Liquid
Gprasp2
GPRASP2 Fusion Protein Ag2800
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documentos
Certificados