missing translation for 'onlineSavingsMsg'
Learn More

Golgin 97 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-81753AF488

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214298

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Golgin 97 Polyclonal antibody specifically detects Golgin 97 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones

Especificaciones

Golgin 97
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 488
gap junction protein, alpha 4, 37kD, golgi autoantigen, golgin subfamily a, 1, golgin A1, Golgin subfamily A member 1, Golgin-97, MGC33154
Golgin 97 antibody was raised against a 17 amino acid peptide near the amino terminus of human Golgin 97. The immunogen is located within amino acids 30 - 80 of Golgin 97. Amino Acid Squence: DFASDGSSSREDLSSQL
0.1 mL
Golgi Apparatus Markers
2800
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales