missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanopartículas de oro, 300 nm, suministradas en 0,1 mg/ml de citrato de sodio con estabilizador, Thermo Scientific Chemicals
Click to view available options
Cantidad:
25 mL
100 mL
Description
For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Para la adsorción de ligandos tiolados como oligonucleótidos y glicoles de polietileno
- Además, debido a la mayor estabilidad, estas nanopartículas de oro son ideales para su uso en aplicaciones físicas y ópticas
Specifications
Nombre del producto químico o material | Gold nanoparticles |
Color | Rosa a rojo |
Nombre de nota | supplied in 0.1mg/mL sodium citrate with stabilizer |
Cantidad | 25 mL |
Olor | Odorless |
Tamaño de partículas | 300nm |
Sensibilidad | Light sensitive |
Formulario | Líquido |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'