missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Feline IgG F(ab')2 Secondary Antibody, FITC, Novus Biologicals™

Goat Polyclonal Antibody

Marca:  Novus Biologicals NBP1-73274

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214112

  • 260.25€ / 2 miligramos
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Feline samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Especificaciones

Especificaciones

IgG F(ab')2
Polyclonal
Fluorescein
immunoglobulin heavy constant gamma 1 (G1m marker)
Cat IgG F(ab')2 fragment
2 mg
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C in the dark.
Flow Cytometry, Immunocytochemistry, Immunofluorescence
LYOPH
Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Goat
Affinity Purified
RUO
No reaction was observed against Cat IgG F(c).
Feline
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only