missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Equine IgG F(ab')2 Secondary Antibody, Biotin, Novus Biologicals™

Goat Polyclonal Antibody

Marca:  Novus Biologicals NBP1-73473

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215235

  • 328.29€ / 2 miligramos
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Especificaciones

Especificaciones

IgG F(ab')2
Polyclonal
Biotin
immunoglobulin heavy constant gamma 1 (G1m marker)
Horse IgG F(ab')2 fragment
2 mg
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Goat
Affinity Purified
RUO
No reaction was observed against Horse IgG F(c).
Equine
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only