missing translation for 'onlineSavingsMsg'
Learn More

GNL1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 14078-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16812564

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Especificaciones

Especificaciones

GNL1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P36915
GNL1, GTP binding protein HSR1, HSR1
Rabbit
IgG
20 μL
-20°C
Primary
2794
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
GNL1
Liquid
GNL1
GNL1 Fusion Protein Ag5203
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados