missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMCL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15575-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
In WB test, the antibody recognizes a 55 kDa band.
This gene encodes a nuclear envelope protein that appears to be involved in spermatogenesis, either directly or by influencing genes that play a more direct role in the process. This multi-exon locus is the homolog of the mouse and drosophila germ cell-less gene but the human genome also contains a single-exon locus on chromosome 5 that contains an open reading frame capable of encoding a highly-related protein.Especificaciones
GMCL1 | |
Polyclonal | |
Unconjugated | |
GMCL1 | |
BTBD13, GCL, GCL1, Germ cell less protein like 1, GMCL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23885, 312516, 64395 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q920G9, Q96IK5 | |
GMCL1 | |
GMCL1 Fusion Protein Ag7929 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |