Learn More
Abcam Glutathione Reductase (GR) Assay Kit
Product Code. 32111475 Shop All Abcam Products
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
Product Code. Quantity Unit Size
32111475 200 Tests 200 tests
1 options
This item is not returnable. View return policy
Product Code. 32111475 Supplier Abcam Supplier No. ab83461200Test
 Restricted Item
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Measure Glutathione Reductase activity in 40m in Biofluids, Cell/Tissue Extracts with Glutathione Reductase (GR) Assay Kit AB83461. Sensitivity >0.1 mU/mL. Cited in 43 publications. Use with Microplate reader.
Measure Glutathione Reductase activity in 40m in Biofluids, Cell/Tissue Extracts with Glutathione Reductase (GR) Assay Kit AB83461. Sensitivity >0.1 mU/mL. Cited in 43 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 200 Tests
Regulatory Status RUO