Learn More
Abcam Glucose-6-Phosphate Dehydrogenase Activity Assay Kit
Product Code. 32112996 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32112996 100 Tests 100 tests
1 options
This item is not returnable. View return policy
Product Code. 32112996 Supplier Abcam Supplier No. ab303735100Test
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Assay Glucose-6-Phosphate Dehydrogenase Activity Assay in 60m in Cell/Tissue Extracts with Glucose-6-Phosphate Dehydrogenase Activity Assay Kit AB303735. Sensitivity ~ 1 μU. Use with Microplate reader.
Assay Glucose-6-Phosphate Dehydrogenase Activity Assay in 60m in Cell/Tissue Extracts with Glucose-6-Phosphate Dehydrogenase Activity Assay Kit AB303735. Sensitivity ~ 1 μU. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO