Learn More
Invitrogen™ Human Glucocorticoid Receptor Recombinant Protein
Glucocorticoid receptor recombinant protein is an insect cell-expressed, full-length, untagged, human nuclear hormone receptor that is partially purified in order to maintain high ligand binding activity. Further purification decreases activity due to loss of endogenous accessory proteins.
Marca:  Invitrogen™ A15663
Detalles adicionales : Peso : 0.01000kg
Código de producto. 15490704
  • 453.00€ / 8.30 miligramo
Fecha estimada de envío: 16-04-2024
para ver el stock
Descripción
Descripción
Studies have shown that glucocorticoid receptor (GR) must be associated with a complex of chaperone proteins for ligand activation. GR binds to known steroids such as dexamethasone with nanomolar affinity.
  • Flexible—use in your in-house assay or as part of our PolarScreen™ Glucocorticoid Receptor Competitor assays for a complete assay solution
  • Ready to use—active and native (untagged)
  • Convenient—available in bulk sizes
  • Used in—PolarScreen™ Glucocorticoid Receptor Competitor Assays (Red and Green)
Especificaciones
Especificaciones
NP_000167
8.3 mg
NR3C1
Recombinant
Store in ultra-cold freezer (-68 to -85°C).
Growth Factors & Receptors
Liquid
2908.0
GR, GCR, GRL, GCCR
Human
Untagged
Baculovirus
Recombinant, Full Length