Learn More
Varilla de carbono vítreo, 3 mm (0,1 in) de diámetro, tipo 1, Thermo Scientific Chemicals
Se emplea como material de electrodo en electroquímica. También se emplea como material de electrodo para la fabricación de sensores. Sirve como material ideal en la tecnología de vacío. También se usa en técnicas de fundición y metalurgia, y en aleaciones duras y crisoles de alta temperatura.
Marca: Thermo Scientific Alfa Aesar 042822.DM
Detalles adicionales : CAS : 7440-44-0 Peso : 0.01000kg
Descripción
Glassy carbon rod is used as an electrode material in electrochemistry. It is also employed as an electrode material for the fabrication of sensors. It serves as an ideal material for the vacuum technology. It is also used in casting, smelting of metal, hard alloys and high temperature crucibles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Glassy carbon rod | |
Tubo | |
100 mm |
7440-44-0 | |
MFCD00133992 | |
Insoluble in water. |
RUO – Research Use Only