Learn More
Heidolph™ Glassware Set - Vertical Condenser (Standard Distillations)
Product Code. 10005711 Shop All Heidolph Instruments Products
Change view
Click to view available options
Electrical Requirements:
230/240 V 50/60 Hz
Unit Size:
Each
Product Code. Electrical Requirements Unit Size
10005711 230/240 V 50/60 Hz Each
1 options
This item is not returnable. View return policy
Product Code. 10005711 Supplier Heidolph™ Supplier No. 5130030000
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Vertical condenser sets have the smallest space requirement and are good for smaller spaces
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
TRUSTED_SUSTAINABILITY
Specifications
Electrical Requirements 230/240 V 50/60 Hz
Voltage 115/230 VAC
Type Evaporation Glassware
Material Borosilicate Glass